missing translation for 'onlineSavingsMsg'
Learn More

Diethylthiocarbamoyl chloride, Thermo Scientific Chemicals

Product Code. 16667052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16667052 25 g 25g
16657052 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 16667052 Proveedor Thermo Scientific Acros N.º de proveedor 465920250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-11-9
Molecular Formula C5H10ClNS
Molecular Weight (g/mol) 151.65
MDL Number MFCD00022106
InChI Key HUUSTUALCPTCGJ-UHFFFAOYSA-N
IUPAC Name (chloromethanethioyl)diethylamine
SMILES CCN(CC)C(Cl)=S

Specifications

Alpha Vector DIETHYLCARBAMOTHIOICCHLORIDE
Melting Point 45°C to 50°C
Color White to Yellow
Boiling Point 113°C
Flash Point 113°C
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 25 g
UN Number 3261
Sensitivity Moisture sensitive
Formula Weight 151.66
Physical Form Solid
Chemical Name or Material Diethylthiocarbamoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.