missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIEXF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
369.00€ - 529.00€
Specifications
| Antigen | DIEXF |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18485451
|
Novus Biologicals
NBP1-88480-25ul |
25 μL |
369.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18235506
|
Novus Biologicals
NBP1-88480 |
0.1 mL |
529.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DIEXF Polyclonal specifically detects DIEXF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| DIEXF | |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 27042 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LLPLDSHGVDFSRVRMWSLNNWSKYYRQTLLFGALQDAQINSVFNKYCVNMQGQVAVRNVPMTGSISHVLVQLPHVFQRMEAENLASVIDARFNFFVNKILPQYRDAVM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C1orf107, chromosome 1 open reading frame 107, DEFFLJ41680, digestive organ expansion factor homolog, digestive organ expansion factor homolog (zebrafish), digestive-organ expansion factor homolog, DJ434O14.5, DKFZp686E19106, FLJ12704, MGC29875 | |
| DIEXF | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title