missing translation for 'onlineSavingsMsg'
Learn More

Difluoro Atorvastatin Calcium Salt, TRC

Product Code. 30006046
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30006046 1 mg 1mg
30005639 10 mg 10mg
30004310 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006046 Supplier Toronto Research Chemicals Supplier No. TRCD4456451MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693793-53-2
Molecular Formula 2 C33 H33 F2 N2 O5 . Ca
Molecular Weight (g/mol) 1191.32
Synonym 1H-Pyrrole-1-heptanoic acid, 2,3-bis(4-fluorophenyl)-β,δ-dihydroxy-5-(1-methylethyl)-4-[(phenylamino)carbonyl]-, calcium salt (2:1), (βR,δR)-, Atorvastatin Related Compound C (USP), (3R,5R)-7-[2,3-Bis(4-fluorophenyl)-5-(1-methylethyl)-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoic acid calcium salt, Fluoroatorvastatin calcium salt
IUPAC Name calcium;(3R,5R)-7-[2,3-bis(4-fluorophenyl)-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
SMILES [Ca+2].CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccc(F)cc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)[O-].CC(C)c5c(C(=O)Nc6ccccc6)c(c7ccc(F)cc7)c(c8ccc(F)cc8)n5CC[C@@H](O)C[C@@H](O)CC(=O)[O-]

Specifications

Quantity 1 mg
InChI Formula InChI=1S/2C33H34F2N2O5.Ca/c2*1-20(2)31-30(33(42)36-25-6-4-3-5-7-25)29(21-8-12-23(34)13-9-21)32(22-10-14-24(35)15-11-22)37(31)17-16-26(38)18-27(39)19-28(40)41;/h2*3-15,20,26-27,38-39H,16-19H2,1-2H3,(H,36,42)(H,40,41);/q;;+2/p-2/t2*26-,27-;/m11./s1
Formula Weight 1190.4341
Chemical Name or Material Difluoro impurity, or (3R,5R)-7-[3-(phenylcarbamoyl)-4,5-bis(4-fluorophenyl)-2-isopropyl-1H-pyrrol-1-yl]-3,5-dihydroxyheptanoic acid calcium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.