missing translation for 'onlineSavingsMsg'
Learn More

Digoxigenin Monodigitoxoside, TRC

Product Code. 18081540
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18081540 10 mg 10mg
18032450 1 mg 1mg
18031450 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18081540 Supplier Toronto Research Chemicals Supplier No. TRCD44650510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5352-63-6
Molecular Formula C29H44O8
Molecular Weight (g/mol) 520.65
Synonym 5β-Card-20(22)-enolide, 3β-(digitoxosyloxy)-12β,14-dihydroxy- (7CI), 5β-Card-20(22)-enolide, 3β-[(2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12β,14-dihydroxy- (8CI), Digoxigenin digitoxoside (6CI), (3β,5β,12β)-3-[(2,6-Dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxycard-20(22)-enolide, D-ribo-Hexopyranoside, digoxigenin-3 2,6-dideoxy-, β- (8CI), Digoxigenin 3-monodigitoxide, Digoxigenin monodigitoxide, Digoxigenin monodigitoxoside
SMILES C[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]2CC[C@@]3(C)[C@H](CC[C@@H]4[C@@H]3C[C@@H](O)[C@]5(C)[C@H](CC[C@]45O)C6=CC(=O)OC6)C2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C29H44O8/c1-15-26(33)22(30)13-25(36-15)37-18-6-8-27(2)17(11-18)4-5-20-21(27)12-23(31)28(3)19(7-9-29(20,28)34)16-10-24(32)35-14-16/h10,15,17-23,25-26,30-31,33-34H,4-9,11-14H2,1-3H3/t15-,17-,18+,19-,20-,21+,22+,23-,25+,26-,27+,28+,29+/m1/s1
Chemical Name or Material Digoxigenin Monodigitoxoside
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.