missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Diguanosine Triphosphate, sodium [G(5')ppp(5')G]

Product Code. 10627075 Shop All Cytiva Products
Change view
Click to view available options
Format:
Solid
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format unitSize
10627075 Solid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627075 Supplier Cytiva Supplier No. 27464301

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Diguanosine Triphosphate, sodium [G(5')ppp(5')G]

TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Content And Storage -20°C
Color White
For Use With (Application) Sample preparation
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.