Learn More
Dihydrostreptomycin sesquisulfate 100 μg/mL in Acetonitrile/Water, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Dihydrostreptomycin sulfate |
| CAS | 5490-27-7 |
| Analyte or Component Names | Dihydrostreptomycin sulfate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C21 H41 N7 O12 . 3 H2 O4 S |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Dihydrostreptomycin sulfate, bis[N,N'''-[(1R,2R,3S,4R,5R,6S)-4-[[5-deoxy-2-O-[2-deoxy-2-(methylamino)-α-l-glucopyranosyl]-3-C-(hydroxymethyl)-α-l-lyxofuranosyl]oxy]-2,5,6-trihydroxycyclohexane-1,3-diyl]diguanidine] trisulfate, D-Streptamine, O-2-deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-(hydroxymethyl)-α-L-lyxofuranosyl-(1→4)-N1,N3-bis(aminoiminomethyl)-, sulfate (2:3), D-Streptamine, O-2-deoxy-2-(methylamino)-α-L-glucopyranosyl-(1→2)-O-5-deoxy-3-C-(hydroxymethyl)-α-L-lyxofuranosyl-(1→4)-N,N'-bis(aminoiminomethyl)-, sulfate (2:3) (salt) (9CI), Streptomycin, dihydro-, sulfate (2:3) (salt) (8CI), Didromycin, Didromycine, Dihydrostreptomycin 3/2 sulfate, Dihydrostreptomycin sesquisulfate, Double-mycin, Panstreptin, Sol-Mycin, Streptomagna |
| InChI Formula | InChI=1S/2C21H41N7O12.3H2O4S/c2*1-5-21(36,4-30)16(40-17-9(26-2)13(34)10(31)6(3-29)38-17)18(37-5)39-15-8(28-20(24)25)11(32)7(27-19(22)23)12(33)14(15)35;3*1-5(2,3)4/h2*5-18,26,29-36H,3-4H2,1-2H3,(H4,22,23,27)(H4,24,25,28);3*(H2,1,2,3,4)/t2*5-,6-,7+,8-,9-,10-,11+,12-,13-,14+,15+,16-,17-,18-,21+;;;/m00.../s1 |
| SMILES | CN[C@H]1[C@H](O)[C@@H](O)[C@H](CO)O[C@H]1O[C@H]2[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]3NC(=N)N)O[C@@H](C)[C@]2(O)CO.CN[C@H]4[C@H](O)[C@@H](O)[C@H](CO)O[C@H]4O[C@H]5[C@H](O[C@H]6[C@H](O)[C@@H](O)[C@H](NC(=N)N)[C@@H](O)[C@@H]6NC(=N)N)O[C@@H](C)[C@]5(O)CO.OS(=O)(=O)O.OS(=O)(=O)O.OS(=O)(=O)O |
| Show More |