missing translation for 'onlineSavingsMsg'
Learn More

Diisohomoeugenol, TRC

Product Code. 30385248
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30385248 25 mg
30381460 250 mg
30374146 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385248 Supplier Toronto Research Chemicals Supplier No. TRCD46443525MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4483-47-0
Molecular Formula C22H28O4
Molecular Weight (g/mol) 356.46
Synonym Dimethyl Ether of Diisoeugenol, NSC 10200, 1-(3,4-Dimethoxyphenyl)-3-ethyl-2,3-dihydro-5,6-dimethoxy-2-methyl-1H-indene, 3-Ethyl-5,6-dimethoxy-1- (3,4-dimethoxyphenyl-2 methyl) -indane
SMILES CCC1C(C)C(c2ccc(OC)c(OC)c2)c3cc(OC)c(OC)cc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H28O4/c1-7-15-13(2)22(14-8-9-18(23-3)19(10-14)24-4)17-12-21(26-6)20(25-5)11-16(15)17/h8-13,15,22H,7H2,1-6H3
Formula Weight 356.1988
Chemical Name or Material Diisohomoeugenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.