missing translation for 'onlineSavingsMsg'
Learn More

Dilauroyl Peroxide, TRC

Product Code. 30386605
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Unit Size:
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386605 5 g 5g
30371998 50 g 50g
30372488 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386605 Supplier Toronto Research Chemicals Supplier No. TRCD6795455G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-74-8
Molecular Formula C24H46O4
Molecular Weight (g/mol) 398.63
Synonym Bis(1-oxododecyl) Peroxide, Alperox C, Alperox F, Dodecanoyl Peroxide, LYP 97, LYP 97F, Laurox Q, Laurox S, Laurox W 40, Laurox W 40GD4, Laurydol, Lup LP, Luperox LP, NSC 670, Peroyl L
IUPAC Name dodecanoyl dodecaneperoxoate
SMILES CCCCCCCCCCCC(=O)OOC(=O)CCCCCCCCCCC

Specifications

Quantity 5 g
InChI Formula InChI=1S/C24H46O4/c1-3-5-7-9-11-13-15-17-19-21-23(25)27-28-24(26)22-20-18-16-14-12-10-8-6-4-2/h3-22H2,1-2H3
Formula Weight 398.3396
Chemical Name or Material Dilauroyl Peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.