missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethomorph-d8 (morpholine-d8) (E/Z mixture), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Dimethomorph-d8 |
| CAS | 1346606-71-0 |
| Molecular Formula | C21 D8 H14 Cl N O4 |
| Quantity | 0.005 g |
| Synonym | Dimethomorph D8 (Morpholine D8) |
| InChI Formula | InChI=1 S/C21H22ClNO4/c1-25-19-8-5-16(13-20(19)26-2)18(15-3-6-17(22)7-4-15)14-21(24)23-9-11-27-12-10-23/h3-8,13-14 H,9-12H2,1-2H3/b18-14+/i9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C1([2 H])OC([2 H])([2 H])C([2 H])([2 H])N(C(=O)\C=C(/c2ccc(Cl)cc2)\c3ccc(OC)c(OC)c3)C1([2 H])[2 H] |
| IUPAC Name | (E)-3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-(2,2,3,3,5,5,6,6-octadeuteriomorpholin-4-yl)prop-2-en-1-one |
| Molecular Weight (g/mol) | 395.906 |
| Formula Weight | 395.174 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction