missing translation for 'onlineSavingsMsg'
Learn More

Dimethoxymethane, 98%, Thermo Scientific Chemicals

Catalog Number p-7081880
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
109-87-5
C3H8O2
76.095
MFCD00008495
NKDDWNXOKDWJAK-UHFFFAOYSA-N
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
8020
CHEBI:48341
dimethoxymethane
COCOC
This item is not returnable. View return policy
109-87-5
C3H8O2
76.095
MFCD00008495
NKDDWNXOKDWJAK-UHFFFAOYSA-N
methylal, formal, methane, dimethoxy, formaldehyde dimethyl acetal, dimethyl formal, anesthenyl, 2,4-dioxapentane, bis methoxy methane, methylene dimethyl ether, methoxymethyl methyl ether
8020
CHEBI:48341
dimethoxymethane
COCOC

Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to protect diols as their methylene acetals, using TMS-OTf/2,6-lutidine:

Solubility
Soluble in water 20°C 330g/L.

Notes
Moisture Sensitive. Store away from oxidizing agents, moisture/water and acids. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point -105°C
Density 0.863
Boiling Point 41°C to 42°C
Flash Point −17°C (1°F)
Linear Formula CH3OCH2OCH3
Refractive Index 1.3545
Quantity 100 mL
UN Number UN1234
Beilstein 1697025
Sensitivity Moisture sensitive
Merck Index 14,6012
Solubility Information Soluble in water 20°C 330g/L.
Formula Weight 76.1
Percent Purity 98%
Chemical Name or Material Dimethoxymethane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'