missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate (Nitrosophenylpyridine Analogue of Nifedipine), Mikromol™

Product Code. 17991356
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17991356 25 mg 25mg
17942086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17991356 Supplier Mikromol Supplier No. MM0003.010025
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Dimethyl 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate (Nitrosophenylpyridine Analogue of Nifedipine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl-2,6-Dimethyl-4-(2-Nitrosophenyl) Pyridine-3,5-Dicarboxylate
CAS 50428-14-3
Analyte or Component Names Dimethyl-2,6-Dimethyl-4-(2-Nitrosophenyl) Pyridine-3,5-Dicarboxylate
Shipping Condition Room Temperature
Molecular Formula C17 H16 N2 O5
Certifications/Compliance ISO 17025
Synonym Nifedipine Imp. B (EP), Nifedipine Nitrosophenylpyridine Analog, Dimethyl 2,6-Dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate, Nitrosophenylpyridine Analogue of Nifedipine, Nifedipine Impurity B
InChI Formula InChI=1S/C17H16N2O5/c1-9-13(16(20)23-3)15(11-7-5-6-8-12(11)19-22)14(10(2)18-9)17(21)24-4/h5-8H,1-4H3
SMILES COC(=O)c1c(C)nc(C)c(C(=O)OC)c1c2ccccc2N=O
IUPAC Name dimethyl 2,6-dimethyl-4-(2-nitrosophenyl)pyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 328.32
Quantity 25 mg
Formula Weight 328.1059 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.