missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Acetylenedicarboxylate, TRC

Product Code. 30385289
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385289 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385289 Supplier Toronto Research Chemicals Supplier No. TRCD46153050G
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl Acetylenedicarboxylate
CAS 762-42-5
Molecular Formula C6H6O4
Quantity 50 g
Synonym 2-Butynedioic Acid 1,4-Dimethyl Ester, 2-Butynedioic Acid Dimethyl Ester, Acetylenedicarboxylic Acid Dimethyl Ester, 1,2-Bis(methoxycarbonyl)acetylene, 1,2-Bis(methoxycarbonyl)ethyne, Bis(carbomethoxy)acetylene, Bis(methoxycarbonyl)acetylene, Butynedioic Acid Dimethyl Ester, Di(carbomethoxy)acetylene, Dimethyl 1,2-Acetylenedicarboxylate, Dimethyl 2-Butynedioate, Dimethyl Butyne-1,4-dioate, Dimethyl Butynedioate, Dimethyl Ethynedicarboxylate, NSC 101119, NSC 14912
InChI Formula InChI=1S/C6H6O4/c1-9-5(7)3-4-6(8)10-2/h1-2H3
SMILES COC(=O)C#CC(=O)OC
IUPAC Name dimethyl but-2-ynedioate
Molecular Weight (g/mol) 142.11
Formula Weight 142.0266
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.