missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Azelate, TRC

Product Code. 30383506
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383506 10 g
30376360 100 g
30386861 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383506 Supplier Toronto Research Chemicals Supplier No. TRCD47063510G
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1732-10-1
Molecular Formula C11H20O4
Molecular Weight (g/mol) 216.27
Synonym Azelaic Acid Dimethyl Ester, Nonanedioic Acid Dimethyl Ester, Dimethyl Azelate, Dimethyl Nonane-1,9-dioate, Dimethyl Nonanedioate, Methyl Azelate, NSC 59040
IUPAC Name dimethyl nonanedioate
SMILES COC(=O)CCCCCCCC(=O)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C11H20O4/c1-14-10(12)8-6-4-3-5-7-9-11(13)15-2/h3-9H2,1-2H3
Formula Weight 216.1362
Chemical Name or Material Dimethyl Azelate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.