missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl cis-stilbene-4,4'-dicarboxylate

Product Code. 11347586
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347586 250 mg 250mg
11337586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347586 Supplier Thermo Scientific Alfa Aesar Supplier No. L11142.MD
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143130-82-9 | C18H16O4 | 296.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143130-82-9
Molecular Formula C18H16O4
Molecular Weight (g/mol) 296.322
MDL Number MFCD00082669
InChI Key JOODVYOWCWQPMV-ARJAWSKDSA-N
Synonym dimethyl cis-stilbene-4,4'-dicarboxylate, dimethyl cis-stilbene-4,4/'-dicarboxylate, z-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, z-4,4'-stilbenedicarboxylic acid dimethyl ester, methyl 4-z-2-4-methoxycarbonylphenyl ethenyl benzoate, methyl 4-z-2-4-methoxycarbonyl phenyl ethenyl benzoate
PubChem CID 7556518
IUPAC Name methyl 4-[(Z)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 112°C to 114°C
Quantity 250 mg
Beilstein 2386934
Formula Weight 296.33
Chemical Name or Material Dimethyl cis-stilbene-4,4'-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.