Learn More
Dimethyl naphthalene-2,6-dicarboxylate, 99+%
Product Code. 11338636
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11338636 5 g 5g
11348636 25 g 25g
11358636 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11338636 Supplier Thermo Scientific Alfa Aesar Supplier No. L17689.06
 Restricted Item
In Stock
Add to basket
missing translation for 'validMainframeMsg'
CAS: 840-65-3 | C14H12O4 | 244.246 g/mol
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.
Solubility
Soluble in hot toluene (very faint turbidity). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 840-65-3
Molecular Formula C14H12O4
Molecular Weight (g/mol) 244.246
MDL Number MFCD00004100
InChI Key GYUVMLBYMPKZAZ-UHFFFAOYSA-N
Synonym dimethyl 2,6-naphthalenedicarboxylate, 2,6-dicarbomethoxynaphthalene, 2,6-naphthalenedicarboxylic acid, dimethyl ester, unii-x4q5t8rs0g, dimethyl-2,6-naphthalenedicarboxylate, 2,6-naphthalenedicarboxylic acid dimethyl ester, x4q5t8rs0g, 2,6-naphthalene dicarboxylic acid, dimethyl ester, dimethylnaphthalene-2,6-dicarboxylate, 2,6-naphthalenedicarboxylic acid, 2,6-dimethyl ester
PubChem CID 61225
IUPAC Name dimethyl naphthalene-2,6-dicarboxylate
SMILES COC(=O)C1=CC2=C(C=C1)C=C(C=C2)C(=O)OC
Specifications
Melting Point 187°C to 193°C
Flash Point 232°C (449°F)
Quantity 5 g
Beilstein 987259
Solubility Information Soluble in hot toluene (very faint turbidity). Insoluble in water.
Formula Weight 244.25
Percent Purity ≥99%
Chemical Name or Material Dimethyl naphthalene-2,6-dicarboxylate