Product Code. 10365890
Shop All Fisher Chemical Products
Click to view available options
Packaging:
Plastic-coated Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
10365890
2.5 L
Plastic-coated Glass Bottle
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Product Code.
10365890
Supplier
Fisher Chemical
Supplier No.
D/4128/PB17
This item is currently unavailable or has been discontinued. View the product page for possible alternatives.
View alternative products
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.
Specifications
Melting Point
18.4°C
CAS
67-68-5
Color
Colorless
Quantity
2.5 L
Packaging
Plastic-coated Glass Bottle
Physical Form
Liquid
Molecular Formula
C2H6OS
Vapor Pressure
0.55mbar at 20°C
MDL Number
MFCD00002089
Viscosity
1.98 mPaS at 25°C
Synonym
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
Boiling Point
189°C
InChI Key
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES
CS(C)=O
IUPAC Name
methylsulfinylmethane
Molecular Weight (g/mol)
78.13
PubChem CID
679
ChEBI
CHEBI:28262
Formula Weight
78.13g/mol
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?