missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl trans-stilbene-4,4'-dicarboxylate, 98+%

Product Code. 11398525
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398525 250 mg 250mg
11388525 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398525 Supplier Thermo Scientific Alfa Aesar Supplier No. L11969.MD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 34541-73-6 | C18H16O4 | 296.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34541-73-6
Molecular Formula C18H16O4
Molecular Weight (g/mol) 296.322
MDL Number MFCD00039527
InChI Key JOODVYOWCWQPMV-ONEGZZNKSA-N
Synonym e-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, dimethyl 4,4'-stilbenedicarboxylate, dimethyl trans-stilbene-4,4'-dicarboxylate, dimethyl stilbene-4,4'-dicarboxylate, benzoic acid, 4,4'-1e-1,2-ethenediylbis-, dimethyl ester, 4,4'-bis carbomethoxy stilbene, methyl 4-1e-2-4-methoxycarbonyl phenyl vinyl benzoate, methyl 4-e-2-4-methoxycarbonyl phenyl ethenyl benzoate, dimethyl 4,4/'-stilbenedicarboxylate
PubChem CID 5378468
IUPAC Name methyl 4-[(E)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 234°C to 236°C
Quantity 250 mg
Beilstein 2471189
Formula Weight 296.33
Percent Purity ≥98%
Chemical Name or Material Dimethyl trans-stilbene-4,4'-dicarboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.