Learn More
Dimethyldimyristylammonium Chloride, TRC
Product Code. 30133629
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133629 2.5 g 2.5g
17944865 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30133629 Supplier Toronto Research Chemicals Supplier No. TRCD4725602.5G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10108-91-5
Molecular Formula C30 H64 N . Cl
Molecular Weight (g/mol) 474.29
Synonym Dimethylditetradecylammonium Chloride, 1-Tetradecanaminium, N,N-dimethyl-N-tetradecyl-, chloride (1:1), 1-Tetradecanaminium, N,N-dimethyl-N-tetradecyl-, chloride (9CI), Ammonium, dimethylditetradecyl-, chloride (8CI), Dimethylditetradecylammonium chloride (7CI), Dimethyldimyristylammonium chloride, Dimyristyl dimethyl ammonium chloride, Ditetradecyldimethylammonium chloride, N,N-Dimethyl-N,N-ditetradecylammonium chloride, Dimethylditetradecylammonium Chloride, 1-Tetradecanaminium, N,N-dimethyl-N-tetradecyl-, chloride (1:1), 1-Tetradecanaminium, N,N-dimethyl-N-tetradecyl-, chloride (9CI), Ammonium, dimethylditetradecyl-, chloride (8CI), Dimethylditetradecylammonium chloride (7CI), Dimyristyl dimethyl ammonium chloride, Ditetradecyldimethylammonium chloride, N,N-Dimethyl-N,N-ditetradecylammonium chloride
IUPAC Name dimethyl-di(tetradecyl)azanium;chloride
SMILES [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCC
Specifications
Quantity 2.5 g
InChI Formula InChI=1S/C30H64N.ClH/c1-5-7-9-11-13-15-17-19-21-23-25-27-29-31(3,4)30-28-26-24-22-20-18-16-14-12-10-8-6-2;/h5-30H2,1-4H3;1H/q+1;/p-1
Formula Weight 473.4727
Percent Purity >95%
Chemical Name or Material Dimethyldimyristylammonium Chloride
Grade HPLC