missing translation for 'onlineSavingsMsg'
Learn More

Dimethyldioctadecyl Ammonium Bromide, 100%, MP Biomedicals™

Product Code. 11496712 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496712 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496712 Supplier MP Biomedicals Supplier No. 0215093905
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Reported to be hightly active agent for enhancement of the immune response. Acts as a substrate for Freund's Adjuvant. Soluble in warm methanol

Specifications

CAS 3700-67-2
Color White
Molecular Formula C38H80BrN
MDL Number MFCD00041975
Quantity 5 g
Synonym dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1
InChI Key PSLWZOIUBRXAQW-UHFFFAOYSA-M
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-]
IUPAC Name dimethyl(dioctadecyl)azanium;bromide
Molecular Weight (g/mol) 630.969
PubChem CID 77293
Formula Weight 630.96
Physical Form Powder
Chemical Name or Material Dimethyldioctadecyl Ammonium Bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.