missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, Certified AR for Analysis, Fisher Chemical

Product Code. 10686252 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10686252 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10686252 Supplier Fisher Chemical Supplier No. D/3900/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 95-45-4
Melting Point 239°C
Color White
Packaging Plastic powder jar
Molecular Formula C4H8N2O2
MDL Number MFCD00002117
Quantity 100 g
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
SMILES C\C(=N/O)\C(\C)=N\O
IUPAC Name N-[(E)-3-nitrosobut-2-en-2-yl]hydroxylamine
Molecular Weight (g/mol) 116.12
PubChem CID 5356010
Formula Weight 116.12g/mol
Physical Form Solid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dimethylglyoxime
Signal Word
  • Danger
Hazard Category
  • Flammable solid Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.