missing translation for 'onlineSavingsMsg'
Learn More

Dimetridazole, TRC

Product Code. 30385837
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385837 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385837 Supplier Toronto Research Chemicals Supplier No. TRCD4796605G
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimetridazole
CAS 551-92-8
Molecular Formula C5 H7 N3 O2
Quantity 5 g
Synonym Dimetridazole, 1H-Imidazole, 1,2-dimethyl-5-nitro-, Imidazole, 1,2-dimethyl-5-nitro- (7CI,8CI), 1,2-Dimethyl-5-nitro-1H-imidazole, 1,2-Dimethyl-5-nitroimidazole, 5-Nitro-1,2-dimethylimidazole, 8595RP, Dimetridazol, Dimetridazole, Dimetrizadole, Dimetronidazole, Emtryl, Emtrylvet, Emtrymix, NSC 226253
InChI Formula InChI=1S/C5H7N3O2/c1-4-6-3-5(7(4)2)8(9)10/h3H,1-2H3
SMILES Cc1ncc([N+](=O)[O-])n1C
IUPAC Name 1,2-dimethyl-5-nitroimidazole
Molecular Weight (g/mol) 141.13
Formula Weight 141.0538
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.