missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DIMT1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97740-50ul
This item is not returnable.
View return policy
Description
DIMT1L Polyclonal antibody specifically detects DIMT1L in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| DIMT1L | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| 18S rRNA (adenine(1779)-N(6)/adenine(1780)-N(6))-dimethyltransferase, DIM1, DIM1 dimethyladenosine transferase 1-like, DIM1 dimethyladenosine transferase 1-like (S. cerevisiae), DIMT1dimethyladenosine transferase, EC 2.1.1, EC 2.1.1.14, EC 2.1.1.183,18S rRNA dimethylase, HSA9761, HUSSY5, N'-adenosyl(rRNA) dimethyltransferase, probable dimethyladenosine transferase, S-adenosylmethionine-6-N' | |
| Produced in rabbits immunized with E. coli-derived Human DIMT1L fragment. | |
| 50 μg | |
| DNA replication Transcription Translation and Splicing | |
| 27292 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction