missing translation for 'onlineSavingsMsg'
Learn More

Dinotefuran UF, TRC

Product Code. 30378902
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30378902 50 mg
30374223 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378902 Supplier Toronto Research Chemicals Supplier No. TRCD48461550MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 457614-34-5
Molecular Formula C7 H14 N2 O2
Molecular Weight (g/mol) 158.2
Synonym Urea, N-methyl-N'-[(tetrahydro-3-furanyl)methyl]-, N-Methyl-N'-[(tetrahydro-3-furanyl)methyl]urea
IUPAC Name 1-methyl-3-(oxolan-3-ylmethyl)urea
SMILES CNC(=O)NCC1CCOC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H14N2O2/c1-8-7(10)9-4-6-2-3-11-5-6/h6H,2-5H2,1H3,(H2,8,9,10)
Formula Weight 158.106
Percent Purity >95%
Chemical Name or Material Dinotefuran-urea
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.