Learn More
DIO2 Mouse anti-Human, Unconjugated, Clone: 1F2A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66813-1-IG-150UL
 View more versions of this product
Product Code. 16839035
  • 410.00€ / 150µL
Description
The DIO2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DIO2. This antibody recognizes human antigen. The DIO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
DIO2
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1F2A1
Unconjugated
150 μL
1734