Learn More
DIO2 Mouse anti-Human, Unconjugated, Clone: 1F2A1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66813-1-IG-150UL
Description
The DIO2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DIO2. This antibody recognizes human antigen. The DIO2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| DIO2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human |
| Western Blot, Immunohistochemistry, ELISA | |
| 1F2A1 | |
| Unconjugated | |
| 150 μL | |
| 1734 | |
| IgG2a |