missing translation for 'onlineSavingsMsg'
Learn More

Dioctyl phthalate, 99%, ACROS Organics™

Product Code. 10154813 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10154813 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10154813 Supplier Thermo Scientific Acros Supplier No. 117091000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Specifications

CAS 117-81-7
Density 0.9810g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C24H38O4
Linear Formula C6H4[COOCH2CH(C2H5)(CH2)3CH3]2
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
MDL Number MFCD00009493
Quantity 100 g
Beilstein 09, IV, 3180
Merck Index 15, 2868
Synonym dehp, bis 2-ethylhexyl phthalate, di 2-ethylhexyl phthalate, diethylhexyl phthalate, di-sec-octyl phthalate, octyl phthalate, fleximel, octoil
InChI Key BJQHLKABXJIVAM-UHFFFAOYSA-N
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1C(=O)OCC(CC)CCCC
IUPAC Name bis(2-ethylhexyl) benzene-1,2-dicarboxylate
Molecular Weight (g/mol) 390.564
PubChem CID 8343
ChEBI CHEBI:17747
Formula Weight 390.55
Vapor Pressure 1.8mbar at 200°C
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Dioctyl phthalate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.