missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphacinone-d4 (indanedione-4,5,6,7-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Diphacinone indanedione-4,5,6,7-d4 |
| CAS | 1219802-15-9 |
| Molecular Formula | C23 2H4 H12 O3 |
| Quantity | 0.01 g |
| Synonym | Diphacinone D4 (indanedione-4,5,6,7 D4) |
| InChI Formula | InChI=1 S/C23H16O3/c24-21-17-13-7-8-14-18(17)22(25)20(21)23(26)19(15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,19-20 H/i7D,8 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2C(=O)C(C(=O)C(c3ccccc3)c4ccccc4)C(=O)c2c1[2 H] |
| IUPAC Name | 4,5,6,7-tetradeuterio-2-(2,2-diphenylacetyl)indene-1,3-dione |
| Molecular Weight (g/mol) | 344.4 |
| Formula Weight | 344.1351 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction