missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Diphenhydramine.HCl, Lipomed
Beskrivelse
Diphenhydramine.HCl
Tekniske data
Tekniske data
| Chemical Name or Material | Diphenhydramine hydrochloride |
| CAS | 147-24-0 |
| Analyte or Component Names | Diphenhydramine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H21 N O . Cl H |
| Synonym | Orphenadrine Hydrochloride Imp. D (EP), Orphenadrine Imp. D (EP), Diphenhydramine Hydrochloride, Orphenadrine Hydrochloride Imp. D (EP) as Hydrochloride, Ethanamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (1:1), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI), Ethylamine, 2-(diphenylmethoxy)-N,N-dimethyl-, hydrochloride (8 CI) |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CN(C)CCOC(c1ccccc1)c2ccccc2 |
| IUPAC Name | 2-benzhydryloxy-N,N-dimethylethanamine;hydrochloride |
| Industry Type | Pharmaceutical |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?