missing translation for 'onlineSavingsMsg'
Learn More

Diphenyl p-Hydroxyphenyl Phosphate, TRC

Product Code. 30010019
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010019 500 mg 500mg
30008435 100 mg 100mg
30008983 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010019 Supplier Toronto Research Chemicals Supplier No. TRCD492418500MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56806-74-7
Molecular Formula C18H15O5P
Molecular Weight (g/mol) 342.28
Synonym Hydroquinonyldiphenyl Phosphate, Diphenyl p-Hydroxyphenylphosphoric Acid Ester;
SMILES Oc1ccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H15O5P/c19-15-11-13-18(14-12-15)23-24(20,21-16-7-3-1-4-8-16)22-17-9-5-2-6-10-17/h1-14,19H
Formula Weight 342.0657
Percent Purity >95%
Chemical Name or Material Diphenyl p-Hydroxyphenyl Phosphate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.