missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-phenyl ester D4 |
| CAS | 1398065-61-6 |
| Molecular Formula | C202H4H10O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzenedicarboxylic acid-3,4,5,6-d4, 1,2-diphenyl ester |
| InChI Formula | InChI=1 S/C20H14O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h1-14 H/i7D,8 D,13 D,14 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)Oc2ccccc2)c(C(=O)Oc3ccccc3)c1[2 H] |
| IUPAC Name | diphenyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 322.35(2) |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction