missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetyl Chloride, TRC

Product Code. 30387457
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387457 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387457 Supplier Toronto Research Chemicals Supplier No. TRCD48699525G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diphenylacetyl Chloride
CAS 1871-76-7
Molecular Formula C14H11ClO
Quantity 25 g
Synonym Diphenylacetyl Acetyl Chloride, Diphenylacetic acid chloride, Diphenylacetyl ChlorideNSC 120906, alpha,alpha-Diphenylacetyl Chloride, alpha-Phenylbenzeneacetyl Chloride
InChI Formula InChI=1S/C14H11ClO/c15-14(16)13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H
SMILES ClC(=O)C(c1ccccc1)c2ccccc2
IUPAC Name 2,2-diphenylacetyl chloride
Molecular Weight (g/mol) 230.69
Formula Weight 230.0498
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.