missing translation for 'onlineSavingsMsg'
Learn More

Diphenylacetylene, 99%

Product Code. 11441738
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441738 100 g 100g
11421738 5 g 5g
11431738 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441738 Supplier Thermo Scientific Alfa Aesar Supplier No. A13979.22
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 501-65-5 | C14H10 | 178.234 g/mol

Diphenylacetylene is often used as a building block in organic and organometallic chemistry. It acts as dienophile and under goes Diels-Alder reaction with tetraphenylcyclopentadienone to prepare hexaphenylbenzene. It acts as a precursor in the preparation of 3-alkoxycyclopropene by reacting with benzal chloride in the presence of potassium t-butoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diphenylacetylene is often used as a building block in organic and organometallic chemistry. It acts as dienophile and under goes Diels-Alder reaction with tetraphenylcyclopentadienone to prepare hexaphenylbenzene. It acts as a precursor in the preparation of 3-alkoxycyclopropene by reacting with benzal chloride in the presence of potassium t-butoxide.

Solubility
Miscible with ether and hot alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 501-65-5
Molecular Formula C14H10
Molecular Weight (g/mol) 178.234
MDL Number MFCD00004786
InChI Key JRXXLCKWQFKACW-UHFFFAOYSA-N
Synonym diphenylacetylene, tolan, diphenylethyne, 1,2-diphenylethyne, tolane, 1,2-diphenylacetylene, benzene, 1,1'-1,2-ethynediyl bis, biphenylacetylene, diphenyl acetylene, ethyne, diphenyl
PubChem CID 10390
ChEBI CHEBI:51579
IUPAC Name 2-phenylethynylbenzene
SMILES C1=CC=C(C=C1)C#CC2=CC=CC=C2

Specifications

Melting Point 58°C to 62°C
Density 0.99
Boiling Point 170°C (19 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 606478
Merck Index 14,9504
Solubility Information Miscible with ether and hot alcohol. Immiscible with water.
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Diphenylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.