missing translation for 'onlineSavingsMsg'
Learn More

Diphenylaminesulfonic Acid Sodium Salt, For ACS analysis, MP Biomedicals™

Product Code. 11442832 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442832 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11442832 Supplier MP Biomedicals Supplier No. 0215251905
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Soluble in water

TRUSTED_SUSTAINABILITY

Specifications

CAS 6152-67-6
Color Beige-Gray
Molecular Formula C12H11NNaO3S
MDL Number MFCD00036468
Quantity 5 g
Synonym sodium 4-phenylamino benzenesulfonate, sodium diphenylamine-4-sulfonate, benzenesulfonic acid, 4-phenylamino-, monosodium salt, unii-3e2ux1382o, sodium 4-anilinobenzenesulfonate, diphenylamine-4-sulfonic acid sodium salt, sodium diphenylamine-p-sulfonate, n-phenylsulfanilic acid, sodium salt, 4-phenylamino benzenesulfonic acid sodium salt, diphenylaminesulfonic acid sodium salt
InChI Key VMVSDVIYIMNZBS-UHFFFAOYSA-N
SMILES [Na].OS(=O)(=O)C1=CC=C(NC2=CC=CC=C2)C=C1
IUPAC Name 4-(phenylamino)benzene-1-sulfonic acid sodium
Molecular Weight (g/mol) 272.27
PubChem CID 517002
Formula Weight 271.27
Physical Form Fine Powder
Chemical Name or Material Diphenylaminesulfonic Acid Sodium Salt, For ACS analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.