missing translation for 'onlineSavingsMsg'
Learn More

Diphenylphosphine, 95%, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10492314 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10492314 50 mL AcroSeal™ Glass bottle 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10492314 Supplier Thermo Scientific Acros Supplier No. 427290500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 829-85-6
Density 1.0700 g/mL
Boiling Point 280.0°C
Flash Point 110°C
Assay Percent Range 95%
Packaging AcroSeal™ Glass bottle
Molecular Formula C12H11P
Linear Formula C6H5PHC6H5
Refractive Index 1.6240 to 1.6260
Quantity 50 mL
Merck Index 15, 3364
Specific Gravity 1.07
Solubility Information Solubility aq. soln.: insoluble.
Molecular Weight (g/mol) 186.19
Formula Weight 186.19
Chemical Name or Material Diphenylphosphine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.