missing translation for 'onlineSavingsMsg'
Learn More

Dipivefrine Hydrochloride, TRC

Product Code. 30377627
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30377627 1 g
30374292 250 mg
30375426 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377627 Supplier Toronto Research Chemicals Supplier No. TRCD4921551G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64019-93-8
Molecular Formula C19 H29 N O5 . Cl H
Molecular Weight (g/mol) 387.9
Synonym Dipivefrine hydrochloride, 4-[(1RS)-1-hydroxy-2-(methylamino)ethyl]-1,2-phenylene bis(2,2-dimethylpropanoate) hydrochloride, Propanoic acid, 2,2-dimethyl-, 1,1'-[4-[1-hydroxy-2-(methylamino)ethyl]-1,2-phenylene] ester, hydrochloride (1:1), Propanoic acid, 2,2-dimethyl-, 4-[1-hydroxy-2-(methylamino)ethyl]-1,2-phenylene ester, hydrochloride (9CI), Propanoic acid, 2,2-dimethyl-, 4-[1-hydroxy-2-(methylamino)ethyl]-1,2-phenylene ester, hydrochloride, (±)-, D Epifrin, Diopine, Diphemin, Dipivefrin hydrochloride, Dipivefrine hydrochloride, Pivalephrine, Propine
IUPAC Name [2-(2,2-dimethylpropanoyloxy)-4-[1-hydroxy-2-(methylamino)ethyl]phenyl] 2,2-dimethylpropanoate;hydrochloride
SMILES Cl.CNCC(O)c1ccc(OC(=O)C(C)(C)C)c(OC(=O)C(C)(C)C)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C19H29NO5.ClH/c1-18(2,3)16(22)24-14-9-8-12(13(21)11-20-7)10-15(14)25-17(23)19(4,5)6;/h8-10,13,20-21H,11H2,1-7H3;1H
Formula Weight 387.1813
Chemical Name or Material Dipivefrine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.