Learn More
Dipropyl Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Dipropyl Sulfate |
| CAS | 598-05-0 |
| Molecular Formula | C6H14O4S |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C6H14O4S/c1-3-5-9-11(7,8)10-6-4-2/h3-6H2,1-2H3 |
| SMILES | CCCOS(=O)(=O)OCCC |
| IUPAC Name | dipropyl sulfate |
| Molecular Weight (g/mol) | 182.24 |
| Formula Weight | 182.0613 |