missing translation for 'onlineSavingsMsg'
Learn More

Dipyridamole, Thermo Scientific™

An inhibitor of adenosine transport and phosphodiesterase

Brand:  Thermo Scientific Alfa Aesar J63329.06

Quantity 5g

 View more versions of this product

Product Code. 15485589

  • ONLINE EXCLUSIVE PRICE
    52.10€
    46.65€ / 5g
    Save 5.45€
    10% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Dipyridamole prevents platelets sticking to the replacement heart valve and causing a blood clot on the valve. It is used to dilate blood vessels in people with peripheral arterial disease and coronary artery disease. This compound has been shown to suppress high glucose-induced osteopontin mRNA expression and protein secretion, as well as inhibit cAMP and cGMP hydrolysis. Research indicates that Dipyridamole is a non-specific nucleoside transport inhibitor with the ability to increase the effects of adenosine in sinoatrial and atrioventricular nodes. Dipyridamole is an inhibitor of ENT1 and ENT2.

Solubility
Soluble in chloroform, methanol, dilute acids, ethanol. Slightly soluble in water.

Notes
Store in a cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

58-32-2
504.636
IZEKFCXSFNUWAM-UHFFFAOYSA-N
3108
2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
C24H40N8O4
MFCD00010555
dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil
CHEBI:4653
C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO

Specifications

58-32-2
MFCD00010555
14,3346
Soluble in chloroform,methanol,dilute acids,ethanol. Slightly soluble in water.
C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO
504.636
CHEBI:4653
Dipyridamole
C24H40N8O4
5g
dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil
IZEKFCXSFNUWAM-UHFFFAOYSA-N
2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol
3108
504.63
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.