Learn More
Dipyridamole, Thermo Scientific™
An inhibitor of adenosine transport and phosphodiesterase
46.65€ - 155.00€
Chemical Identifiers
| CAS | 58-32-2 |
|---|---|
| Molecular Formula | C24H40N8O4 |
| Molecular Weight (g/mol) | 504.636 |
| MDL Number | MFCD00010555 |
| InChI Key | IZEKFCXSFNUWAM-UHFFFAOYSA-N |
| Synonym | dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil |
| PubChem CID | 3108 |
| ChEBI | CHEBI:4653 |
| IUPAC Name | 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol |
| SMILES | C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15485589
|
Thermo Scientific Alfa Aesar
J63329.06 |
5g |
52.10€ 46.65€ / 5g Save 5.45€ 10% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15495589
|
Thermo Scientific Alfa Aesar
J63329.14 |
25g |
155.00€
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsDipyridamole prevents platelets sticking to the replacement heart valve and causing a blood clot on the valve. It is used to dilate blood vessels in people with peripheral arterial disease and coronary artery disease. This compound has been shown to suppress high glucose-induced osteopontin mRNA expression and protein secretion, as well as inhibit cAMP and cGMP hydrolysis. Research indicates that Dipyridamole is a non-specific nucleoside transport inhibitor with the ability to increase the effects of adenosine in sinoatrial and atrioventricular nodes. Dipyridamole is an inhibitor of ENT1 and ENT2.
Solubility
Soluble in chloroform, methanol, dilute acids, ethanol. Slightly soluble in water.
Notes
Store in a cool dry place. Keep away from oxidizing agents.
Chemical Identifiers
| 58-32-2 | |
| 504.636 | |
| IZEKFCXSFNUWAM-UHFFFAOYSA-N | |
| 3108 | |
| 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol |
| C24H40N8O4 | |
| MFCD00010555 | |
| dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil | |
| CHEBI:4653 | |
| C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO |
Specifications
| 58-32-2 | |
| MFCD00010555 | |
| dipyridamole, dipyridamine, persantin, dipyridamol, dipyudamine, persantine, curantyl, stimolcardio, cardoxin, kurantil | |
| IZEKFCXSFNUWAM-UHFFFAOYSA-N | |
| 2-[[2-[bis(2-hydroxyethyl)amino]-4,8-di(piperidin-1-yl)pyrimido[5,4-d]pyrimidin-6-yl]-(2-hydroxyethyl)amino]ethanol | |
| 3108 | |
| 504.63 |
| C24H40N8O4 | |
| 14,3346 | |
| Soluble in chloroform,methanol,dilute acids,ethanol. Slightly soluble in water. | |
| C1CCN(CC1)C2=NC(=NC3=C2N=C(N=C3N4CCCCC4)N(CCO)CCO)N(CCO)CCO | |
| 504.636 | |
| CHEBI:4653 | |
| Dipyridamole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 200-374-7
missing translation for 'rtecsNumber' : KK7450000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.