Learn More
B Braun™ Discofix™ C 3-Way Stopcocks
Product Code. 10166673
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10166673 Blue Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10166673 Supplier B Braun™ Supplier No. 16494C
  • ONLINE EXCLUSIVE PRICE
    187.46€
    164.13€ / Pack of 50
    Save 23.33€
    12% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
A stopcock system for infusion therapy and monitoring.
  • Resistant to the effects of pharmaceuticals with a previously unattained degree of transparency
  • Pressure resistant up to 2 bar
  • Free of latex, PVC and DEHP
  • Resistance against stress cracking resulting from exposure to infusion solutions, drugs and alcohol-based disinfectants
  • Protection against the danger of unnoticed cracks, air infusion and leakage
  • Fast connection with rotating adapter
  • Reliable, secure system connection with rotation of the connected stopcock unit on its own axis
  • Light snap at every 45° turn of the valve
  • Allows convenient access for injections and aspirations by rotating adapter
  • Lets you feel the proper setting by tactile response
  • Applications include parenteral nutrition therapy, anesthesia with IV, chemotherapy with cytostatic agents, immunotherapy and neurology
Specifications
Color Blue
For Use With (Application) Infusion therapy and monitoring
Latex Free Yes
Product Type Stopcock
Max. Pressure 2 bar
Quantity 50/Pk.