missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DISP1 Monoclonal Antibody (768213)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561320

Product Code. 30535907

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.

The pattern of cellular proliferation and differentiation that leads to normal development of embryonic structures often depends upon the localized production of secreted protein signals. Cells surrounding the source of a particular signal respond in a graded manner according to the effective concentration of the signal, and this response produces the pattern of cell types constituting the mature structure. A novel segment-polarity gene known as dispatched has been identified in Drosophila and its protein product is required for normal Hedgehog signaling. This gene is one of two human homologs of Drosophila dispatched and, based on sequence identity to its mouse counterpart, the encoded protein may play an essential role in Hh patterning activities in the early embryo.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

DISP1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q96F81
DISP1
E. coli-derived recombinant human DISP1, Glu212-Lys492 and Ser746-Thr984.
100 μg
Primary
Human
Antibody
IgG2b
Immunocytochemistry
768213
Unconjugated
DISP1
1190008H24Rik; chameleon; con; Disp; Disp1; Dispa; dispatched 1; dispatched A; dispatched homolog 1; dispatched homolog 1 (Drosophila); dispatched RND transporter family member 1; DKFZP434I0428; FLJ43740; helix; hlx; hypothetical protein LOC619201; Icb; Icbins; Mdispa; MGC104180; MGC13130; MGC16796; multipass transmembrane protein; Disp1; protein chameleon; Protein dispatched homolog 1; protein dispatched homolog 1 pseudogene; twelve-pass transmembrane protein; zgc:111866
Mouse
Protein A/G
RUO
84976
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.