missing translation for 'onlineSavingsMsg'
Learn More

Disperse Yellow 64 (Technical Grade), TRC

Product Code. 30004171
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30004171 500 mg 500mg
30134080 1 g 1g
30009342 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004171 Supplier Toronto Research Chemicals Supplier No. TRCD494840500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10319-14-9
Molecular Formula C18 H10 Br N O3
Molecular Weight (g/mol) 368.18
Synonym 1,3-Indandione, 2-(4-bromo-3-hydroxy-2-quinolyl)- (7CI,8CI), 2-(4-Bromo-3-hydroxy-2-quinolinyl)-1H-indene-1,3(2H)-dione, 3'-Hydroxy-4'-bromoquinophthalone, 4-Bromo-3-hydroxy-2-(1,3-indandion-2-yl)quinoline, 4-Bromo-3-hydroxyquinophthalone, Allilon Yellow 3G, C.I. 47023, C.I. Disperse Yellow 64, C.I. Solvent Yellow 176, Dianix Yellow E 3G, Dianix Yellow F 3G-E, Dianix Yellow F 3G-E concentrate, Disperse Yellow 3G, Disperse Yellow 4GL, Disperse Yellow 64, Disperse Yellow E 3G, Dispersion Yellow VP 247, Miketon Polyester Yellow 3GSL, Navilene Yellow 3G, Palanil Yellow 3G, Resolin Yellow 3GL, Samaron Yellow H 3GL, Solvaperm Yellow G, Solvent Yellow 176, Sumikaron Yellow SE 3GL, Tersetile Yellow 3GL, Transetile Yellow P 3GL, Tulasteron Fast Yellow 3G-B
IUPAC Name 2-(4-bromo-3-hydroxyquinolin-2-yl)indene-1,3-dione
SMILES Oc1c(Br)c2ccccc2nc1C3C(=O)c4ccccc4C3=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H10BrNO3/c19-14-11-7-3-4-8-12(11)20-15(18(14)23)13-16(21)9-5-1-2-6-10(9)17(13)22/h1-8,13,23H
Formula Weight 366.9844
Chemical Name or Material Disperse Yellow 64
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.