missing translation for 'onlineSavingsMsg'
Learn More

Disperse Yellow 9 (Technical Grade), TRC

Product Code. 30010790
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010790 2.5 g 2.5g
30007743 1 g 1g
30004496 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010790 Supplier Toronto Research Chemicals Supplier No. TRCD9953602.5G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6373-73-5
Molecular Formula C12 H10 N4 O4
Molecular Weight (g/mol) 274.23
Synonym 1,4-Benzenediamine, N-(2,4-dinitrophenyl)- (9CI), p-Phenylenediamine, N-(2,4-dinitrophenyl)- (6CI,7CI,8CI), N1-(2,4-Dinitrophenyl)-1,4-benzenediamine, 2,4-Dinitro-4'-aminodiphenylamine, 4-Amino-2',4'-dinitrodiphenylamine, C.I. 10375, C.I. Disperse Yellow 9, C.I. Solvent Orange 53, Disperse Yellow 9, N-(2,4-Dinitrophenyl)-1,4-phenylenediamine, NSC 60560, SRA Golden Yellow IX, Seriplas Yellow PL, Serisol Fast Yellow PL, Serisol Fast Yellow PL 150, Solvent Orange 53, Supracet Yellow 5RD
IUPAC Name 1-N-(2,4-dinitrophenyl)benzene-1,4-diamine
SMILES Nc1ccc(Nc2ccc(cc2[N+](=O)[O-])[N+](=O)[O-])cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C12H10N4O4/c13-8-1-3-9(4-2-8)14-11-6-5-10(15(17)18)7-12(11)16(19)20/h1-7,14H,13H2
Formula Weight 274.0702
Chemical Name or Material Disperse Yellow 9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.