Learn More
Copan Italia™ Disposable Inoculation Loops
Product Code. 11746538
Change view
Click to view available options
Color:
Dark Blue
Light Blue
Quantity:
2000/Pk.
4000/Pk.
500/Pk.
Unit Size:
Pack of 2000
Pack of 4000
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
11746538 Light Blue 500/Pk. Pack of 500
13414992 Dark Blue 4000/Pk. Pack of 4000
11756538 Light Blue 2000/Pk. Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11746538 Supplier Copan Italia™ Supplier No. 179CS01
Edge
Use Copan Italia™ Disposable Inoculation Loops for research and laboratory needs. The versatile inoculation loop is made of rigid plastic, and is calibrated and sterile.
  • Sterile
  • Calibrated
  • Rigid plastic
Specifications
Disposable Single-use
Material Plastic
Sterility Sterile
Color Light Blue
Product Type Inoculating Loop
For Use With (Equipment) Laboratory and Research use
Quantity 500/Pk.
For Use With (Application) Laboratory and Research use