Learn More
Copan Italia™ Disposable Inoculation Loops
Product Code. 13414992
Change view
Click to view available options
Color:
Dark Blue
Light Blue
Quantity:
2000/Pk.
4000/Pk.
500/Pk.
Unit Size:
Pack of 2000
Pack of 4000
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity Unit Size
13414992 Dark Blue 4000/Pk. Pack of 4000
11746538 Light Blue 500/Pk. Pack of 500
11756538 Light Blue 2000/Pk. Pack of 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13414992 Supplier Copan Italia™ Supplier No. 8177CSR40H
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Use Copan Italia™ Disposable Inoculation Loops for research and laboratory needs. The versatile inoculation loop is made of rigid plastic, and is calibrated and sterile.
  • Sterile
  • Calibrated
  • Rigid plastic
Specifications
Disposable Single-use
Volume 10 μL
Material Rigid Plastic
Sterility Sterile
Color Dark Blue
Product Type Inoculating Loop
For Use With (Equipment) Laboratory and Research use
Quantity 4000/Pk.
Calibrated Yes
For Use With (Application) Laboratory and Research use