Learn More
Copan Italia™ Disposable Pasteur Pipets
Product Code. 11705769
Change view
Click to view available options
Volume (Metric):
6 mL
7 mL
Bulb Draw (Metric):
3.5 mL
Unit Size:
Pack of 400
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Bulb Draw (Metric) Unit Size
11705769 6 mL 3.5 mL Pack of 400
11775759 7 mL 3.5 mL Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11705769 Supplier Copan Italia™ Supplier No. 204C
  • ONLINE EXCLUSIVE PRICE
    101.64€
    33.87€ / Pack of 400
    Save 67.77€
    66% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Use Copan Italia™ Disposable Pasteur Pipets for biological fluids and most acids. Their low surface affinity reduces the loss of cells and proteins of interest.
  • Unbreakable
  • Can be sealed and frozen
  • Usable in liquid nitrogen
  • Sterile
  • Polyethylene LDPE
Specifications
Bulb Draw (Metric) 3.5 mL
Diameter (Metric) Outer 5 mm
Material LDPE
Drops/mL 22
For Use With (Application) For transferring and dispensing liquids safely in all types of laboratories
Graduated Yes
Quantity 400 Pack
Volume (Metric) 6 mL
Length (Metric) 230 mm
Type Pasteur Pipette
Disposable Single-use