Learn More
Scientific Industries SI™ Disruptor Genie™ Digital Cell Disruptor
Product Code. 15577345 Shop All Ohaus Products
Change view
Click to view available options
Plug Type:
EU Plug
Model:
SI-DD58
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Model Unit Size
15577345 EU Plug SI-DD58 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15577345 Supplier Scientific Industries SI™ Supplier No. 83041287
  • ONLINE EXCLUSIVE PRICE
    819.00€
    785.80€ / Pack of 1
    Save 33.20€
    4% Off
View all promotions
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Increases the disruption efficiency of disruption beads with unique, patented microtube holder design. Scientific Industries SI™ Disruptor Genie™ Digital Cell Disruptor provides vigorous mechanical cell disruption.
    • Digital model is ideal for accurate, reproducible, repeatable results.
    • Features adjustable speed and speed alarm
  • Rapid hands-free disruption of up to 12 microtubes (1.5mL or 2.0mL)
  • Supplied with NEW Universal Microtube Holder
  • Set timer to run for up to 15 minutes or continuously
  • Suitable for use in cold rooms or incubators
  • Less costly than ultrasonic units or homogenizers
Applications include:
  • Disruption of bacterial cultures for nucleic acid isolation and downstream processing
  • Grinding plant tissue for DNA extractions
  • Preparation of cell lysates from yeast or fungi
  • Rapid resuspension of pellets
  • Dissolution of chemical substances
TRUSTED_SUSTAINABILITY
Specifications
Speed Range 1000 to 2850 rpm
Electrical Requirements 230 V, 50 Hz
Voltage 230 V
Holds Microtubes
Tube Capacity 1.5/2 mL
Plug Type EU Plug
Frequency 50 Hz
Depth (Metric) 165 mm
Height (Metric) 190 mm
Width (Metric) 122 mm
Weight (Metric) 4.3 kg
Model SI-DD58
Includes Universal microtube holder
No. of Positions 12