Learn More
Hanna Instruments™ Dissolved Oxygen Electrolyte Solutions
Product Code. 10643272
Change view
Click to view available options
For Use With (Equipment):
HI Galvanic DO Probes
HI Polarographic DO Probes
Unit Size:
30mL
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10643272 HI Polarographic DO Probes 30mL
11534347 HI Galvanic DO Probes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10643272 Supplier Hanna Instruments™ Supplier No. HI7041S
  • ONLINE EXCLUSIVE PRICE
    34.08€
    24.32€ / 30mL
    Save 9.76€
    28% Off
View all promotions
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Capacity (Metric) 30 mL
For Use With (Equipment) HI Polarographic DO Probes
Type Dissolved Oxygen Electrolyte Solution