Learn More
Hanna Instruments™ DiST™ 3 EC Tester
Product Code. 10708401
Change view
Click to view available options
Range (Conductivity):
0 to 1999 mS/cm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range (Conductivity) Unit Size
10708401 0 to 1999 mS/cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Product Code. 10708401 Supplier Hanna Instruments™ Supplier No. HI98303
  • ONLINE EXCLUSIVE PRICE
    95.81€
    72.94€ / Pack of 1
    Save 22.87€
    23% Off
View all promotions
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
R\u00E9cupel-bijdrage uitgesloten
Regulatory: This product is under eco-contribution tax.
Specifications
Range (Conductivity) 0 to 1999 mS/cm
Resolution (Conductivity) 1 μS/cm
Type EC Tester
Accuracy ±2% Full-scale