missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FKBP10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29566
This item is not returnable.
View return policy
Description
FKBP10 Polyclonal antibody specifically detects FKBP10 in Human,Mouse samples. It is validated for Western Blot
Specifications
| FKBP10 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 60681 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 65 kDa FK506-binding protein, 65 kDa FKBP, EC 5.2.1.8, FK506 binding protein 10 (65 kDa), FK506 binding protein 10, 65 kDa, FK506-binding protein 10, FKBP-10, FKBP6, FKBP-65, FKBP65PPIase FKBP10, FLJ20683, FLJ22041, FLJ23833, hFKBP65, Immunophilin FKBP65, OI6, peptidyl-prolyl cis-trans isomerase FKBP10, PPIASE, Rotamase | |
| Between 450 and 500 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction