missing translation for 'onlineSavingsMsg'
Learn More

DL-2-Methylbutyric Acid, TRC

Product Code. 30474543
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474543 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474543 Supplier Toronto Research Chemicals Supplier No. TRCM29486525G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methylbutyric acid
CAS 116-53-0
Molecular Formula C5 H10 O2
Quantity 25 g
Synonym 2-Methylbutyric Acid, Butanoic acid, 2-methyl- (9CI, ACI), 2-Methylbutanoic acid (ACI), Butyric acid, 2-methyl- (6CI, 8CI), Butyric acid, α-methyl- (3CI), (±)-2-Methylbutanoic acid, (±)-2-Methylbutyric acid, (±)-Methylethylacetic acid, (±)-α-Methylbutyric acid, 2-Ethylpropanoic acid, 2-Methyl-n-butyric acid, 2-Methylbutyric acid, DL-2-Methylbutanoic acid, DL-2-Methylbutyric acid, DL-α-Methylbutyric acid, Active valeric acid, Ethylmethylacetic acid, Hydroangelic acid, Methylethylacetic acid, NSC 7304, α-Methyl-n-butyric acid, α-Methylbutyric acid
InChI Formula InChI=1S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)
SMILES CCC(C)C(=O)O
IUPAC Name 2-methylbutanoic acid
Molecular Weight (g/mol) 102.13
Formula Weight 102.0681
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.