missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Alanine-2-d1-N-FMOC, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-DL-alanine-2-D |
| Molecular Formula | C18 D H16 N O4 |
| Quantity | 0.25 g |
| Synonym | N-[(9 H-Fluoren-9-ylmethoxy)carbonyl]-DL-alanine-2-D, 2-[[(9 H-Fluoren-9-ylmethoxy)carbonyl]amino]propanoic-2-d acid, FMOC-DL-Alanine-2-d, N-(9-Fluorenylmethoxycarbonyl)-DL-alanine-2-d, N-Fmoc-DL-alanine-2-d, DL-Alanine-2-d1-N-FMOC |
| InChI Formula | InChI=1 S/C18H17NO4/c1-11(17(20)21)19-18(22)23-10-16-14-8-4-2-6-12(14)13-7-3-5-9-15(13)16/h2-9,11,16 H,10H2,1H3,(H,19,22)(H,20,21)/i11D |
| SMILES | [2 H]C(C)(NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| IUPAC Name | 2-deuterio-2-(9 H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
| Molecular Weight (g/mol) | 312.338 |
| Formula Weight | 312.122 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction